Order Entry
Portugal
ContactUsLinkComponent
Supelco® Organic Reference Standard, N-Methylaniline
Supelco® Organic Reference Standard, N-Methylaniline
Catalog # 49636-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, N-Methylaniline
Catalog # 49636-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    100-61-8
  • Description:
    N-Methylaniline
  • Seq:
    2210
  • Storage conditions:
    RT
  • Hazard class description:
    H301 + H311 + H331 - H319 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008283
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Monomethylaniline
  • Melting point:
    –57 °C (lit.)
  • Formula:
    C₆H₅NHCH₃
  • Molecular weight:
    107,15
  • Density:
    0,989 g/ml at 25 °C (lit.)
  • Flash point:
    85 °C - Closed cup
  • Boiling point:
    196 °C (lit.)

Specifications

About this item

  • GHS signal: GHS06, GHS08, GHS09
Caution: P273 - P280 - P301 + P310 - P302 + P352 + P312 - P304 + P340 + P311 - P314