Order Entry
Portugal
ContactUsLinkComponent
Measuring beakers, conical, graduated
Measuring beakers, conical, graduated
Catalog # USBE4410
Supplier #: 4410
undefined
Measuring beakers, conical, graduated
Catalog # USBE4410
Supplier #: 4410
Log in for availability
Select an Option
100 mm
Select an Option
1000 ml
Select an Option
155 mm
Select an Option
1
About this item
Stainless steel, heavy pattern, with handle.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
  • Internal graduation
Ideal for corrosive materials. Robust and durable.
Frequently Bought Together