Order Entry
Portugal
ContactUsLinkComponent
Benzoic acid
Benzoic acid
Catalog # EHERC10537500
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Benzoic acid
Catalog # EHERC10537500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029
  • Environmentally Preferable:

Specifications

About this item

 :
 :122.12 g/mol
Boiling Pt:249
Flash Pt:121 °C (closed cup)
EINECS:200-618-2
REACH:01-2119455536-33
 :GHS07
Storage Temperature:Ambient
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
Merck Index:13,01092