Benzaldehyde for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-52-7
:
Formula:C₆H₅CHO
MDL Number:MFCD00003299
:106.12 g/mol
Merck Index:13,01057
Boiling Pt:179 °C (1013 hPa)
Melting Pt:–26 °C
Density:1.049 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-860-4
UN:1990
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:64 °C
:GHS07
Specification Test Results
- Solubility:3.3 g/l (20 °C)
- Melting Point:-26 °C
- Molar Mass:106.12 g/mol
- Boiling Point:179 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (20 °C)
- Flash Point:64 °C
- Refractive Index:1.545 (20 °C, 589 nm)
- Explosion Limit:1.4 - 8.5 %(V)
- Density:1.05 g/cm3 (20 °C)
- pH:5.9 (1 g/l, H2O)
- Ignition Point:190 °C
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Related Information
Frequently Bought Together