Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dapsone, Supelco®

Dapsone, Supelco®

Glass bottle for solids, 250 mg
Catalog # 46158-250MG
Irritant
Supplier:   Merck
Dapsone, Supelco®
Catalog # 46158-250MG
Supplier:  Merck
CAS Number:  80-08-0
Dapsone, Supelco®
Dapsone, Supelco®

Glass bottle for solids, 250 mg

CAS Number:  80-08-0
Price
52,10 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:201-248-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg

Specifications