2,2,2-Trifluoroethyl acrylate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:407-47-6
:
Formula:C₅H₅F₃O₂
MDL Number:MFCD00000444
:154.09 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:206-987-6
UN:3272
Frequently Bought Together