Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dapsone 97%

Dapsone 97%

Irritant
Dapsone 97%
Certificates
Dapsone 97%

2 Options Available Below

Product Details & Documents
Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Melting Pt:173…178 °C
  • EINECS:201-248-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    173 to 178 °C
  • Titration with HClO4:
    ≥96.0 %
  • Apparent density:
    0.3 to 0.5 g/cm3
  • Titration:
    ≥97.5 % w/w (as Oxalate) On dry substance
  • Iron (Fe):
    ≤200 ppm
  • Insoluble matter:
    ≤0.3 % (in HCl)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 47,60 €
    47,60 €
    1 * 100 g
       47,60 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 121,00 €
    121,00 €
    1 * 500 g
       121,00 €