


Bromobenzene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-86-1
:
Formula:C₆H₅Br
MDL Number:MFCD00000055
:157.01 g/mol
Merck Index:13,01390
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–31 °C
Density:1.495 g/cm³ (20 °C)
EINECS:203-623-8
UN:2514
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:51 °C
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:4 g/l (20 °C)
- Melting Point:-31 °C
- Molar Mass:157 g/mol
- Boiling Point:156 °C (1013 hPa)
- Vapor Pressure:4 hPa (20 °C)
- Flash Point:51 °C
- Refractive Index:1.5602 (20 °C, 589 nm)
- Explosion Limit:0.5 - 2.5 %(V)
- Density:1.49 g/cm3 (20 °C)
- Ignition Point:565 °C
- Saturation concentration:26 g/m3 (20 °C) Air
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together