Order Entry
Portugal
Orders LinkContactUsLinkComponent
Myristic acid 99%

Myristic acid 99%

Myristic acid 99%
Certificates
Myristic acid 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Tetradecylic acid, Tetradecanoic acid
  • CAS Number:544-63-8
  • Formula:C₁₄H₂₈O₂
  • MDL Number:MFCD00002744
  • Molecular Weight:228.38 g/mol
  • Merck Index:12,06416
  • Boiling Pt:326 °C (760 mmHg)
  • Melting Pt:50…57 °C
  • Density:0,8622 g/cm³ (20 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:208-875-2

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder or flakes or chunks or crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    50 to 57 °C
  • GC:
    ≥98.5 %
  • Titration with NaOH:
    ≥98.5 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.