Order Entry
Portugal
ContactUsLinkComponent
 
 
 
Caproic acid for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
Caproic acid for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    9.7 g/l (20 °C)
  • Melting Point:
    -3 °C
  • Molar Mass:
    116.16 g/mol
  • Boiling Point:
    206 °C (1013 hPa)
  • Vapor Pressure:
    0.3 hPa (20 °C)
  • Flash Point:
    110 °C
  • Refractive Index:
    1.4162 (20 °C, 589 nm)
  • Density:
    0.93 g/cm3 (20 °C)
  • pH:
    4 (1 g/l, H2O, 20 °C)
  • Ignition Point:
    380 °C

 

Frequently Bought Together