Caproic acid 99%
:142-62-1
:
:C₆H₁₂O₂
:MFCD00004421
:116.16 g/mol
:13,01765
:202…203 °C (760 mmHg)
:–3 °C
:0.92 g/cm³
:104 °C
:Ambient
:205-550-7
:2829
:8,III
:GHS05
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4150 to 1.4180 (20°C, 589 nm)
- GC:≥98.5 %
- Acid value:473 to 493 mg KOH/g
- test 11:Result 11
- test 12:Result 12
- test 13:Result 13
- test 14:Result 14
- test 15:Result 15
- test 16:Result 16
- test 17:Result 17
- test 18:Result 18
- test 19:Result 19
- test 20:Result 20
- test 21:Result 21
- test 22:Result 22
- test 23:Result 23
- test 24:Result 24
- test 25:Result 25
- test 26:Result 26
- test 27:Result 27
- test 28:Result 28
- test 29:Result 29
- test 30:Result 30
- test 31:Result 31
- test 32:Result 32
- test 33:Result 33
- test 34:Result 34
- test 35:Result 35
- test 36:Result 36
- test 37:Result 37
- test 38:Result 38
- test 39:Result 39
- test 40:Result 40
- test 41:Result 41
- test 42:Result 42
- test 43:Result 43
- test 44:Result 44
- test 45:Result 45
- test 46:Result 46
- test 47:Result 47
- test 48:Result 48
- test 49:Result 49
- test 50:Result 50
- test 51:Result 51
- test 52:Result 52
- test 53:Result 53
- test 54:Result 54
- test 55:Result 55
- test 56:Result 56
- test 57:Result 57
Frequently Bought Together