
Sodium hydroxide 5 mol/l (5 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:1.186 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):4.99 - 5.01 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500)
Informações de Embalagem
31624_BSE TSE - SODIUM HYDROXYDE 5 MOLL 5 N
Certificados BSE/TSE
spec_VWRC31624.290 spec_VWRC31624.320 spec_VWRC31624.320
Especificação do Produto
VWR® Volumetric Solutions for Titration
Literatura do Produto
Frequently Bought Together