Order Entry
Portugal
Orders LinkContactUsLinkComponent
Forceps
Supplier:   Brand
Forceps
Supplier:  Brand
Certificates
Forceps
2 Options Available Below
Product Details & Documents
PMP, white.
  • Pointed ends
  • Autoclavable (121 °C)
Product Family Options
Product Information
  • Type
    Colour
    Length
    Availability
    Price
  • 231-0247
    Tweezers, in PMP
    Tweezers, in PMP
    Straight, pointed
    White
    115 mm
    Log in for availability
    Log in for availability
    1 * 10 unid.
    1 * 10 unid. - 23,10 €
    23,10 €
    1 * 10 unid.
       23,10 €
  • 231-0248
    Tweezers, in PMP
    Tweezers, in PMP
    Straight, pointed
    White
    145 mm
    Log in for availability
    Log in for availability
    1 * 10 unid.
    1 * 10 unid. - 27,00 €
    27,00 €
    1 * 10 unid.
       27,00 €