

Antimony(III) potassium oxitartrate trihydrate ≥99.0%, EMPLURA®, Supelco®
: Merck
:28300-74-5
:304-61-0
:C₈H₄K₂O₁₂Sb₂·3H₂O
:MFCD00167056
:667.88 g/mol
:13,00706
:100 °C
:2.6 g/cm³ (20 °C)
:229-436-1
:6.1,III
:M-Clarity™ quality level = MQ200
:1551
:GHS07
:GHS09
- Assay (iodometric):99.0 - 103.0 %
- Identity:passes test
- As (Arsenic):≤ 0.015 %
- Pb (Lead):≤ 0.002 %
- Loss on drying:≤ 2.7 %
Frequently Bought Together