Order Entry
Portugal
ContactUsLinkComponent
High accuracy reference thermometer, KM20REF
High accuracy reference thermometer, KM20REF
Catalog # 620-1169
Supplier:  COMARK
undefined
High accuracy reference thermometer, KM20REF
Catalog # 620-1169
Supplier:  COMARK

Specifications

  • Range:
    −100…+199,9 °C
  • Resolution:
    0,1 °C
  • Description:
    Ultra-accurate digital thermometer, KM20REF
  • Environmentally Preferable:
  • Accuracy:
    ±0,2 °C
  • Pk:
    1

Specifications

About this item

This highly accurate, electronic reference thermometer with PT100 probe allows you to check the calibration of all types of thermometer and probe combinations.

  • Platinum Resistance Thermometer (PRT) technology and permanently attached PRT probe for highest system accuracy
  • Automatic switch-off

The KM20REF thermometer can be used in-conjunction with the KM820/VAL Validation Calibration Check Unit to calibrate contact and infrared thermometers.

Delivery: Supplied with a hard carrying case, a nine-volt IEC 6LF22 alkaline battery for 100 hours of use and a five-point UKAS certificate of calibration.