Order Entry
Portugal
ContactUsLinkComponent
Cleanroom wipes, on perforated rolls, Durx
Cleanroom wipes, on perforated rolls, Durx
Catalog # 115-0497
Supplier:  BERKSHIRE
undefined
Cleanroom wipes, on perforated rolls, Durx
Catalog # 115-0497
Supplier:  BERKSHIRE
Specifications
  • Packed:
    Perforated roll
  • L×W:
    380×340 mm
  • Pk:
    1 Roll
Specifications
About this item
Non woven dry perforated roll for use in non critical areas. Polyester cellulose blend, for general use in manufacture of devices sensitive to fibre and particle contamination, non volatile residues and ions.
  • Good absorbency for excellent spill control and wet wiping
  • High wet strength
  • No chemical binders
Suitable for use in Class 100, ISO 5, M3.5 cleanrooms.