Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery
Supplier: Brand
Certificates
About this item
AR-Glas®.
- Calibrated to deliver (TD, Ex); type 1: Zero on top
- Marks and inscriptions in high contrast blue enamel
- Includes a batch certificate
- Also available (on request) with an individual certificate, USP individual certificate or DAkks calibration certificate
- Only pipettes marked (*) are suitable for cotton plugs
DIN EN ISO 835
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Certifications: DIN EN ISO 835. DE-M marking.