Cell culture plates, Nunc™ Multidish
Supplier: Thermo Fisher Scientific
Certificates
About this item
PS, sterile, with lid. Useful in all areas of cell culture, including scale up and cloning.
- Raised well rims lower the risk of cross-contamination
- Flat bottom wells allow optimum optical quality
- Nunclon™Δ certified surface treatment
Specifications
- Catalog No:
- 734-2176
- 391-8036
- 734-2157
- 734-2372
- 734-2156
- 734-0991
- 734-0992
- 734-1147
- 734-0993
- 734-2373
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement