Human/Mouse Recombinant TGF-β 3 (from E. coli)
About this item
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
- High Quality
- Low Endotoxin
- Affordable
Shelf life:12 months from date of receipt when stored at 4 °C as supplied.
: For research use only.
4 Options Available Below
Product Details & Documents
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
- High Quality
- Low Endotoxin
- Affordable
Shelf life:12 months from date of receipt when stored at 4 °C as supplied.
: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT800-07-10UG
- SHBT800-07-100UG
- SHBT800-07-500UG
- SHBT800-07-1MG
- Protein/peptide name:
- TGF-β 3
- TGF-β 3
- TGF-β 3
- TGF-β 3
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human|Mouse
- Human|Mouse
- Human|Mouse
- Human|Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P10600/P17125
- P10600/P17125
- P10600/P17125
- P10600/P17125
- Sequence:
- MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
- MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
- MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
- MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
- Amino acid number:
- 113/226
- 113/226
- 113/226
- 113/226
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Form:
- Liquid
- Liquid
- Liquid
- Liquid
- Formulation:
- 10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/ml
- 10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/ml
- 10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/ml
- 10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/ml
- Molecular weight:
- 12,9/25,7 kDa
- 12,9/25,7 kDa
- 12,9/25,7 kDa
- 12,9/25,7 kDa
- Storage conditions:
- Store at 4 °C
- Store at 4 °C
- Store at 4 °C
- Store at 4 °C
- Shipping temperature:
- Ice pack
- Ice pack
- Ice pack
- Ice pack
- Purification:
- Reducing and non-reducing SDS-PAGE analysis
- Reducing and non-reducing SDS-PAGE analysis
- Reducing and non-reducing SDS-PAGE analysis
- Reducing and non-reducing SDS-PAGE analysis
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT800-07-10UGProtein/peptide nameTGF-β 3Protein/peptide typeRecombinantProtein functionCytokineSpeciesHuman|MouseSourceE. coliConjugationUnconjugatedGene IDP10600/P17125SequenceMALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCSAmino acid number113/226Protease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLiquidFormulation10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/mlMolecular weight12,9/25,7 kDaStorage conditionsStore at 4 °CShipping temperatureIce packPurificationReducing and non-reducing SDS-PAGE analysisTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT800-07-100UGProtein/peptide nameTGF-β 3Protein/peptide typeRecombinantProtein functionCytokineSpeciesHuman|MouseSourceE. coliConjugationUnconjugatedGene IDP10600/P17125SequenceMALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCSAmino acid number113/226Protease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLiquidFormulation10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/mlMolecular weight12,9/25,7 kDaStorage conditionsStore at 4 °CShipping temperatureIce packPurificationReducing and non-reducing SDS-PAGE analysisTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT800-07-500UGProtein/peptide nameTGF-β 3Protein/peptide typeRecombinantProtein functionCytokineSpeciesHuman|MouseSourceE. coliConjugationUnconjugatedGene IDP10600/P17125SequenceMALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCSAmino acid number113/226Protease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLiquidFormulation10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/mlMolecular weight12,9/25,7 kDaStorage conditionsStore at 4 °CShipping temperatureIce packPurificationReducing and non-reducing SDS-PAGE analysisTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT800-07-1MGProtein/peptide nameTGF-β 3Protein/peptide typeRecombinantProtein functionCytokineSpeciesHuman|MouseSourceE. coliConjugationUnconjugatedGene IDP10600/P17125SequenceMALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCSAmino acid number113/226Protease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLiquidFormulation10 mM acetic acid and 20% ethanol at a concentration of 0,25 mg/mlMolecular weight12,9/25,7 kDaStorage conditionsStore at 4 °CShipping temperatureIce packPurificationReducing and non-reducing SDS-PAGE analysisTested applicationsBioassay



