Order Entry
Portugal
ContactUsLinkComponent
Sudan II 90% (dye content), Sigma-Aldrich®
Irritant
Sudan II 90% (dye content), Sigma-Aldrich®
Catalog # 199656-25G
CAS Number:  
undefined
Irritant
Sudan II 90% (dye content), Sigma-Aldrich®
Catalog # 199656-25G
CAS Number:  

Specification Test Results

  • Appearance:
    Orange to dark red powder
  • Infrared spectrum:
    Conforms to Structure
  • UV-visible spectrum:
    In methanol (E494 (±3 nm) = 14000 (minimum); E422 (±3 nm) = 9000 (minimum))
  • Carbon:
    68.8 - 87.6%
  • Nitrogen:
    8.9 - 11.4%
  • Solubility:
    10 mg/ml, CHCL₃; clear to opaque, orange to red solution

Specification Test Results

About this item