PowerPlex® 35GY System
PowerPlex® 35GY system, 200 (or 400 direct amplification) reactions
Catalog # PROMDC3520
Supplier: Promega
Jump to:
Product Details & Documents
The PowerPlex® 35GY system is the first 8-dye STR multiplex and allows co-amplification and fluorescent detection of the 20 CODIS core loci plus Amelogenin and DYS391 for sex determination. The inclusion of two additional dye channels relative to previous STR multiplex kits provides improved and more informative analyses for even your most challenging DNA samples.
- Fifteen mini-STRs improve results with degraded samples
- System contains the shortest SE33 amplicon of any STR chemistry kit available
- Penta D, Penta E and SE33 loci allow wider database searching
The PowerPlex® 35GY QI Reagent is required for direct amplification and assists in determining if a sample is degraded or inhibited.
Specifications
- Description:PowerPlex® 35GY system, 200 (or 400 direct amplification) reactions