EasySep™ Human ILC2 Enrichment Kit
STMC17972
- Description:EasySep™ human ILC2 enrichment kit, 1×10⁹ cells
- Environmentally Preferable:
- Pk:1 KIT
Immunomagnetic enrichment of untouched human ILC2 cells.
- Fast, easy-to-use and column-free
- Untouched, viable cells
- Facilitates rapid flow sorting of ILC2s
The EasySep™ Human ILC2 enrichment kit is designed to enrich Group 2 Innate Lymphoid Cells (ILC2s) from leukapheresis products by immunomagnetic negative selection. Unwanted cells are targeted for removal by Tetrameric Antibody Complexes directed against non-ILC2s and dextran-coated magnetic particles (RapidSpheres™). Labeled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube as desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry and cell sorting.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).