3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
PROMH50210001
: Promega
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
:
:614.89 g/mol
:13,02052
:116 °C
:Refrigerator
:75621-03-3
:C₃₂H₅₈N₂O₇S
:MFCD00081083