Order Entry
Portugal
Orders LinkContactUsLinkComponent
PQ401

PQ401

Supplier:   TCI
PQ401
Supplier:  TCI
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  1-(5-Chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea, N-(5-Chloro-2-methoxyphenyl)-N’-(2-methyl-4-quinolinyl)urea, IGF-IR Inhibitor
  • CAS Number:196868-63-0
  • Formula:C₁₈H₁₆ClN₃O₂
  • MDL Number:MFCD00160558
  • Molecular Weight:341.8 g/mol
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 90,00 €
    90,00 €
    1 * 10 mg
       90,00 €

  • 50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 289,00 €
    289,00 €
    1 * 50 mg
       289,00 €