Order Entry
Portugal
ContactUsLinkComponent
 
Anti-Desmin Rabbit Polyclonal Antibody
  ANTIA8809-100
New Product
undefined
Anti-Desmin Rabbit Polyclonal Antibody
  ANTIA8809-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CMD1I
  • Antigen symbol:
    Desmin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17661
  • Antigen synonyms:
    CSM1|FLJ41013|DESM_HUMAN|DES|Desmin|FLJ12025|FLJ39719|FLJ41793|CSM2
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    54 kDa
  • Sequence:
    NYIEKVRFLEQQNAALAAEVNRLKGREPTRVAELYEEELRELRRQVEVLTNQRARVDVERDNLLDDLQRLKAKLQEEIQLKEEAENNLAAFRADVDAATLARIDLERRIESLNEEIAFLKKVHEEEIRELQAQLQEQQVQVEMDMSKPDLTAALRDIRAQYETIAAKNISEAEEWYKSKVSDLTQAANKNNDALRQAKQEMMEYRHQIQSYTCEIDALKGTNDSLMRQMRELEDRFASEASGYQDNIARLEEEIRHLKDEMARHLREYQDLLNVKMALDVEIATYRKLLEGEESRINLPIQTYSALNFRETSPEQRGSEVHTKKTVMIKTIETRDGEVVSEATQQQHEVL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 121-470 of human Desminmin (NP_001918.3)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to Desmin for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:20

Type: Primary
Antigen: Desmin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat