Anti-Desmin Mouse Monoclonal Antibody [clone: D33]
Catalog # ANTIA251515-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CMD1I
- Antigen symbol:Desmin
- Clonality:Monoclonal
- Clone:D33
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Hamster,Chicken,Human,Rat
- Form:Liquid
- Gene ID:UniprotID# P17661
- Antigen synonyms:CSM1|FLJ41013|DESM_HUMAN|DES|Desmin|FLJ12025|FLJ39719|FLJ41793|CSM2
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Proteins from human Leiomyoma
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [D33] antibody to Desmin for IHC-P with samples derived from Human, Rat, Hamster and Chicken.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Desmin
Clonality: Monoclonal
Clone: D33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat, Hamster, Chicken