Order Entry
Portugal
ContactUsLinkComponent
Anti-Desmin Rabbit Monoclonal Antibody [clone: ARC0235]
Anti-Desmin Rabbit Monoclonal Antibody [clone: ARC0235]
Catalog # ANTIA305689-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Desmin Rabbit Monoclonal Antibody [clone: ARC0235]
Catalog # ANTIA305689-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CMD1I
  • Antigen symbol:
    Desmin
  • Clonality:
    Monoclonal
  • Clone:
    ARC0235
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17661
  • Antigen synonyms:
    CSM1|FLJ41013|DESM_HUMAN|DES|Desmin|FLJ12025|FLJ39719|FLJ41793|CSM2
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    54 kDa
  • Sequence:
    EEEIRHLKDEMARHLREYQDLLNVKMALDVEIATYRKLLEGEESRINLPIQTYSALNFRETSPEQRGSEVHTKKTVMIKTIETRDGEVVSEATQQQHEVL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 371 - 470 of human Desmin (P17661).
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC0235] antibody to Desmin for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Desmin
Clonality: Monoclonal
Clone: ARC0235
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat