Order Entry
Portugal
ContactUsLinkComponent
 
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (Alexa Fluor® 488)
 
undefined
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (Alexa Fluor® 488)

 

  • Catalog No:
  • STMC60014AD.1
  • STMC60014AD
  • Antigen name:
  • CD38
  • CD38
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • HIT2
  • HIT2
  • Reactivity:
  • Horse, Chimpanzee, Human
  • Horse, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 952
  • 952
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together