Order Entry
Portugal
ContactUsLinkComponent
Anti-CD24 Rat Monoclonal Antibody [clone: M1/69] (FITC)
Anti-CD24 Rat Monoclonal Antibody [clone: M1/69] (FITC)
Certificates
undefined
Anti-CD24 Rat Monoclonal Antibody [clone: M1/69] (FITC)

Specifications

  • Catalog No:
  • STMC60099FI.1
  • STMC60099FI
  • Antigen name:
  • CD24
  • CD24
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • M1/69
  • M1/69
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12484
  • 12484
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together