Specifications
- Antigen name:CD298
- Clonality:Monoclonal
- Clone:LNH-94
- Conjugation:PE (Phycoerythrin)
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:483
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal IgG1 antibody against human CD298, PE-conjugated.
- B cells
- Granulocytes
- Subsets
- Lymphocytes
- Mammary cells
- Monocytes
- T cells
The LNH-94 antibody reacts with human CD298, the β3 subunit of the Na+/K+ ATPase. The Na+/K+ ATPase is a 42 kDa type II transmembrane protein composed of four catalytic α subunits and three smaller glycosylated non-catalytic β subunits. The Na+/K+ ATPase is important for establishing electrochemical gradients and maintaining normal resting membrane potential by coupling ATP hydrolysis to the active transport of Na+ and K+. The β subunits have been reported to act as chaperones that stabilize the α subunits and occlude K+. CD298 has been used to identify metastatic human cancer cells in mouse xenograft models and has been reported to modulate aspects of immunity, including HIV production, and T cell and B cell activation. CD298 is found on leukocytes, platelets, and most other tissue types.
Type:
Antigen: CD298
Clonality: Monoclonal
Clone: LNH-94
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human