Order Entry
Portugal
ContactUsLinkComponent
Anti-CD117 Rat Monoclonal Antibody [clone: ACK2] (PE)
Anti-CD117 Rat Monoclonal Antibody [clone: ACK2] (PE)
Certificates
undefined
Anti-CD117 Rat Monoclonal Antibody [clone: ACK2] (PE)

Specifications

  • Catalog No:
  • STMC60034PE.1
  • STMC60034PE
  • Antigen symbol:
  • CD117
  • CD117
  • Antigen name:
  • c-KIT
  • c-KIT
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • ACK2
  • ACK2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16590
  • 16590
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together