Order Entry
Portugal
ContactUsLinkComponent
o-Tolidine ≥95%
o-Tolidine ≥95%
Certificates
undefined
o-Tolidine ≥95%
Certificates

Specification Test Results

  • Appearance (Colour):
    White to Light Brown
  • Appearance (Form):
    Powder or Chunks
  • Proton NMR spectrum:
    Conforms to Structure
  • Purity (Titration by HClO4):
    ≥95 %

Specification Test Results

Related Information

Frequently Bought Together