Order Entry
Portugal
ContactUsLinkComponent
Potassium bromide 99,99%, HighPurS-CERT
Potassium bromide 99,99%, HighPurS-CERT
Catalog # ACSDPSPBRC.100G
Supplier:  CPACHEM
New Product
CAS Number:  
Potassium bromide 99,99%, HighPurS-CERT
Catalog # ACSDPSPBRC.100G
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:1435 °C
Melting Pt:734 °C
Density:1,4 g/cm³
 :119 g/mol
EINECS:231-830-3
 :GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358