MethoCult™ M3630 Medium
Catalog # STMC03630
Supplier: STEMCELL TECHNOLOGIES
CAS Number:
Specifications
- Pk:100 mL
- Description:MethoCult™ M3630
Specifications
About this item
Methylcellulose-based medium for mouse pre-B lymphoid progenitor cells.
MethoCult™ M3630 is optimised for the growth and enumeration of mouse pre-B lymphoid progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow and Whitlock-Witte long-term cultures.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').