Order Entry
Portugal
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (mFluor™ Violet 510)
Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (mFluor™ Violet 510)
Catalog # AATB10454110
Supplier:  AAT Bioquest
undefined
Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (mFluor™ Violet 510)
Catalog # AATB10454110
Supplier:  AAT Bioquest

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD45 antigen
  • Antigen symbol:
    CD45
  • Clonality:
    Monoclonal
  • Clone:
    2D1
  • Conjugation:
    mFluor™ Violet 510
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Storage temperature:
    2...8 °C with minimized light exposure. Do not freeze.
  • Shipping temperature:
    20
  • Purification:
    Affinity chromatography
  • Pack type:
    Vial
  • Pk:
    100 Tests

Specifications

About this item

2D1 is an anti-human monoclonal antibody that recognizes the CD45 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2D1
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype: lgG1 kappa
Reactivity: Human