Anti-DCAF4L1 Rabbit Polyclonal Antibody
Catalog # PRSI55-487
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:DDB1- and CUL4-associated factor 4-like protein 1
- Antigen symbol:DCAF4L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino Acid Range = 241-270
- Form:Liquid
- Gene ID:Q3SXM0
- Antigen synonyms:DCAF4L1
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:44 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This WDR21B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 241-270 amino acids from the C-terminal region of human WDR21B.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
The specific function of WDR21B is not yet known.
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: DCAF4L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human