Anti-RUNX2 Rabbit Polyclonal Antibody
Catalog # PRSI33-058
Supplier: ProSci Inc.
Specifications
- Tipo de anticorpo:Primário
- Nome do antigénio:runt-related transcription factor 2
- Símbolo do antígeno:RUNX2
- Clonalidade:Policlonal
- Conjugado:Não conjugado
- Hospedeiro:Rabbit
- Isótipo:IgG
- Reatividade:Human,Mouse
- Western blot:Yes
- Formato:Purified Antibody
- Forma:Liquid
- ID do gene:Q13950
- Sinônimos de antígeno:RUNX2
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Temperatura de armazenamento:Aliquot and Store at –20 °C. Avoid freez-thaw cycles.
- Concentração:0,5 mg/ml
- Shipping temperature:4 °C
- Imunogênio:A recombinant protein fragment from the internal region of human RUNX2 was used as the immunogen for this antibody.
- Purification:Protein A affinity chromatography
- Tamanho:100 µg
- Und.:100 µG
Specifications
About this item
RUNX (runt-related transcription factor) proteins are a family of transcription factors involved in many different biological systems, including cell differentiation and cancer. RUNX2 is an essential factor in bone development, acting as a scaffold protein for nucleic acids and regulatory factors involved in skeletal gene expression. It is also involved in cancer cell movement, activating factors involved in the metastasis of prostate and breast cancer cells to bone.
Western blot: 1-3 µg/ml Provided assay concentrations are sµggestions only, RUNX2 antibody titration may be required for optimal results.
Type: Primary
Antigen: RUNX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse