Order Entry
Portugal
ContactUsLinkComponent
Anti-CAPG Rabbit Polyclonal Antibody
Anti-CAPG Rabbit Polyclonal Antibody
Catalog # PRSI22-864
Supplier:  ProSci Inc.
Anti-CAPG Rabbit Polyclonal Antibody
Catalog # PRSI22-864
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Capping protein (actin filament), gelsolin-like
  • Antigen symbol:
    CAPG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    822
  • Antigen synonyms:
    gelsolin-like capping protein|capping protein (actin filament)|actin regulatory protein CAP-G|macrophage-capping protein|macrophage capping protein|CAPG|MCP|AFCP|gelsolin-like|actin-regulatory protein CAP-G
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-348 of human CAPG (NP_001738.2)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: CAPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat