Anti-RUNX3 Rabbit Polyclonal Antibody
Catalog # PRSI22-848
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Runt-related transcription factor 3
- Antigen symbol:RUNX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:864
- Antigen synonyms:AML2|Core-binding factor subunit alpha-3|RUNX3|PEA2-alpha C|SL3/AKV core-binding factor alpha C subunit|SL3-3 enhancer factor 1 alpha C subunit|PEBP2A3|Runt-related transcription factor 3|Oncogene AML-2|CBFA3|Polyomavirus enhancer-binding protein 2 alpha C subunit|Acute myeloid leukemia 2 protein|PEBP2-alpha C|CBF-alpha-3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 210-429 of human RUNX3 (NP_001026850.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RUNX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human