Anti-PTGES2 Rabbit Polyclonal Antibody
Catalog # PRSI22-732
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Prostaglandin E synthase 2
- Antigen symbol:PTGES2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:80142
- Antigen synonyms:GBF-1|mPGES-2|gamma-interferon-activated transcriptional element-binding factor 1|C9orf15|mPGE synthase-2|prostaglandin E synthase 2|prostaglandin-H(2) E-isomerase|membrane-associated prostaglandin E synthase 2|PGES2|microsomal prostaglandin E synthase-2|GATE-binding factor 1|GBF1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 88-377 of human PTGES2 (NP_079348.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PTGES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat