Anti-MARCKSL1 Rabbit Polyclonal Antibody
PRSI22-727
- Antibody type:Primary
- Antigen name:MARCKS-related protein
- Antigen symbol:MARCKSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:65108
- Antigen synonyms:MARCKS-related protein|MARCKSL1|Mac-MARCKS|MacMARCKS|Macrophage myristoylated alanine-rich C kinase substrate|MARCKS-like protein 1|MLP|MRP
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-195 of human MARCKSL1 (NP_075385.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MARCKSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat