Anti-SPA17 Rabbit Polyclonal Antibody
PRSI19-463
- Antibody type:Primary
- Antigen name:Sperm autoantigenic protein 17
- Antigen symbol:SPA17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:53340
- Antigen synonyms:cancer/testis antigen 22|CT22|sperm protein 17|sperm surface protein Sp17|SP17|SP17-1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-151 of human SPA17 (NP_059121.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SPA17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat