Anti-Ly6g Rat Monoclonal Antibody [clone: 1A8] (FITC (Fluorescein Isothiocyanate))
PRSI77-106
- Antibody type:Primary
- Antigen name:lymphocyte antigen 6 complex, locus G
- Antigen symbol:Ly6g
- Clonality:Monoclonal
- Clone:1A8
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:P35461
- Antigen synonyms:Ly6g
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
The 1A8 monoclonal antibody specifically reacts with the 21 to 25 kDa glycophosphatidylinositol-anchored protein known as Ly-6G, expressed by the granulocytes from the bone marrow and periphery neutrophils. Ly-6G and Ly-6C form the Granulocyte Receptor-1 antigen (GR-1). The binding of the 1A8 antibody to the Ly-6G can be blocked by another antibody, RB6-8C5, which also recognizes Ly-6C. While 1A8 is specific for only Ly-6G, RB6-8C5 also binds to Ly-6C.
Type: Primary
Antigen: Ly6g
Clonality: Monoclonal
Clone: 1A8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse