Anti-Spike Protein Rabbit Recombinant Antibody [clone: 9C1A3]
PRSI11-102
- Antibody type:Primary
- Antigen name:Surface glycoprotein
- Antigen symbol:Spike Protein
- Clonality:Recombinant
- Clone:9C1A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Virus
- Form:Liquid
- Gene ID:43740568
- Antigen synonyms:Spike glycoprotein
- Modification:None
- Storage buffer:Supplied as a 0,2 µm filtered solution in PBS, pH 7,4
- Concentration:1 mg/ml
- Shipping temperature:20
- Immunogen:HEK293 derived SARS-COV-2 SPIKE RBD, amino acids Arg319-Phe541
- Purification:Affinity purified, endotoxin level: <1,0 EU/µg of the protein by LAL method
- Pack type:Vial
- Pk:0,1 mg
Type: Primary
Antigen: Spike protein
Clonality: Recombinant
Clone: 9C1A3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Virus