Anti-Nucleoprotein Rabbit Recombinant Antibody [clone: [85E9]]
PRSI11-021
- Antibody type:Primary
- Antigen name:Nucleocapsid Phosphoprotein
- Antigen symbol:Nucloprotein
- Clonality:Recombinant
- Clone:[85E9]
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG1 kappa
- Reactivity:Virus
- Western blot:Yes
- Form:Liquid
- Gene ID:43740575
- Antigen synonyms:SARS-CoV-2 (COVID-19) Nucleoprotein Antibody [85E9]: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
- Modification:None
- Storage buffer:1X PBS, 0,02% NaN₃
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:SARS-CoV-2 nucleoprotein (NP) specific peptide
- Purification:SARS-CoV-2 (COVID-19) nucleoprotein antibody [85E9] is affinity chromatography purified via peptide column
- Pack type:Vial
- Pk:0,1 mg
Type: Primary
Antigen: Nucleoprotein
Clonality: Recombinant
Clone: [85E9]
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1k
Reactivity: Virus