Anti-ABCC4 Rabbit Polyclonal Antibody
Catalog # PRSI62-908
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Multidrug resistance-associated protein 4
- Antigen symbol:ABCC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:1117-1145 aa
- Form:Liquid
- Gene ID:10257
- Modification:None
- Molecular weight:150 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ABCC4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1117-1145 amino acids from the C-terminal region of human ABCC4.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ABCC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 1117-1145
Host: Rabbit
Isotype: Ig
Reactivity: Human