Anti-DET1 Rabbit Polyclonal Antibody
Catalog # PRSI24-006
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:DET1 partner of COP1 E3 ubiquitin ligase
- Antigen symbol:DET1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:55070
- Antigen synonyms:COP1 ubiquitin ligase partner|de-etiolated homolog 1|DET1|DET1 homolog
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 15-240 of human DET1 (NP_060466.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat