Order Entry
Portugal
ContactUsLinkComponent
Anti-STX Recombinant Antibody [Clone: SP6]
Anti-STX Recombinant Antibody [Clone: SP6]
Catalog # PRSI24-300
Supplier:  ProSci Inc.
Anti-STX Recombinant Antibody [Clone: SP6]
Catalog # PRSI24-300
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Clone:
    SP6
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Hamster,
    Pig,
    Rabbit,
    Human,
    Bovine,
    Guinea pig,
    Dog,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    STX
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: STX
Clonality: Recombinant
Clone: SP6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human;Rat;Hamster;Rabbit;Pig;Dog;Guinea pig;Bovine