Order Entry
Portugal
ContactUsLinkComponent
Anti-CD37 Recombinant Antibody [Clone: WR17]
Anti-CD37 Recombinant Antibody [Clone: WR17]
Catalog # PRSI24-264
Supplier:  ProSci Inc.
Anti-CD37 Recombinant Antibody [Clone: WR17]
Catalog # PRSI24-264
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD37 molecule
  • Antigen symbol:
    CD37
  • Clonality:
    Recombinant
  • Clone:
    WR17
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    951
  • Antigen synonyms:
    GP52-40|TSPAN26
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Chronic lymphocytic leukaemia cells
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: CD37 molecule
Clonality: Recombinant
Clone: WR17
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human