Order Entry
Portugal
ContactUsLinkComponent
Anti-IFNG Recombinant Antibody [Clone: D9D10]
Anti-IFNG Recombinant Antibody [Clone: D9D10]
Catalog # PRSI24-236
Supplier:  ProSci Inc.
Anti-IFNG Recombinant Antibody [Clone: D9D10]
Catalog # PRSI24-236
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HGNC:HGNC:5438
  • Antigen symbol:
    IFNG
  • Clonality:
    Recombinant
  • Clone:
    D9D10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Isotype:
    IgG kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    3458
  • Antigen synonyms:
    IFNG|IFN-gamma|Interferon gamma|Immune interferon
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    FPLC-purified natural human IFN-y. Refer to the original article [PMID: 3114127]
  • Tested applications:
    Blocking
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: HGNC:HGNC:5438
Clonality: Recombinant
Clone: D9D10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human